Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf59 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
SDP

Catalog No. 501672491
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2491 100 μL
50-254-2361 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2491 Supplier OriGene Supplier No. TA501348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The function of the C21orf59 protein remains unknown.

Specifications

Antigen C21orf59
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2E5
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C21orf59
Gene Accession No. P57076
Gene Alias C21orf48, CILD26, FBB18, Kur
Gene Symbols C21orf59
Host Species Mouse
Immunogen Full length human recombit protein of human C21orf59 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56683
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.