Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542790 Supplier Thermo Scientific Supplier No. PA542790
Add to Cart
Add to Cart

C21orf59 Polyclonal Antibody

C21orf59 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf59
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C21ORF59
Gene Accession No. P57076
Gene Alias C21orf48; C21orf59; CFAP298; chromosome 21 open reading frame 59; CILD26; Cilia- and flagella-associated protein 298; FBB18; Kur; prostate cancer upregulated protein 1; Protein kurly homolog; UPF0769 protein C21orf59
Gene Symbols C21ORF59
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C21orf59 (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.