Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C21orf69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569394 Supplier Invitrogen™ Supplier No. PA569394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C21orf69 Polyclonal Antibody for Western Blot

C21orf67 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf69
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LINC01547
Gene Accession No. P58512
Gene Alias C21orf67; C21orf69; LINC01547; long intergenic non-protein coding RNA 1547; PRED54
Gene Symbols LINC01547
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C21orf69.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84536
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.