Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C21orf7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559979 Supplier Invitrogen™ Supplier No. PA559979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISVLCSATSL AMLEDNPKVS KLATGDWMLT LKPKSITVPV EIPSSPLDDT PPEDSIPLVF PELDQQLQPL PPCHDSEESM EVFKQHCQI Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 42%.

C21orf7 plays a critical role in the TGF-beta signaling transduction pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP3K7CL
Gene Accession No. P57077
Gene Alias C1H21orf7; C21orf7; C31H21orf7; HC21ORF7; MAP3K7 C-terminal like; MAP3K7 C-terminal-like protein; Map3k7cl; ORF63; TAK1L; TAK1-like protein; TAK1-like protein 1; TAK1-like protein 2; TAK1-like protein 4; TAKL; TAKL-1; TAKL-2; TAKL-4; TGF-beta activated kinase; TGF-beta-activated kinase like
Gene Symbols MAP3K7CL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C21orf7. Recombinant protein control fragment (Product #RP-91819).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.