Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C21orf7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542201
Description
Peptide sequence: DSEESMEVFK QHCQIAEEYH EVKKEITLLE QRKKELIAKL DQAEKEKVDA Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
C21orf7 plays a critical role in the TGF-beta signaling transduction pathway.
Specifications
| C21orf7 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K7CL | |
| C1H21orf7; C21orf7; C31H21orf7; HC21ORF7; MAP3K7 C-terminal like; MAP3K7 C-terminal-like protein; Map3k7cl; ORF63; TAK1L; TAK1-like protein; TAK1-like protein 1; TAK1-like protein 2; TAK1-like protein 4; TAKL; TAKL-1; TAKL-2; TAKL-4; TGF-beta activated kinase; TGF-beta-activated kinase like | |
| Rabbit | |
| Protein A | |
| RUO | |
| 56911 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P57077 | |
| MAP3K7CL | |
| Synthetic peptide directed towards the C-terminal of human C21orf7 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction