Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C21orf91 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | C21orf91 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
C21orf91 Polyclonal specifically detects C21orf91 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C21orf91 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54149 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CRNSVLWPHSHNQAQKKEETISSPEANVQTQHPHYSREELNSMTLGEVEQLNAKLLQQIQEVFEELTHQVQEKDSLASQLHVRHVAIEQLLKNCSKLPCLQVGRTGMKSHLP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
C21orf14, C21orf38, C21orf7, chromosome 21 open reading frame 38, chromosome 21 open reading frame 91, CSSG1, DKFZp781D1223, early undifferentiated retina and lens, EURL, protein EURL homolog, YG81, YG81C21orf14 | |
C21ORF91 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title