Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C21orf91 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565538 Supplier Invitrogen™ Supplier No. PA565538
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEEQFVNIDLN DDNICSVCKL GTDKETLSFC HICFELNIEG VPKSDLLHTK SLRGHKDCFE KYHLIANQGC PRSKLS Highest antigen sequence identity to the following orthologs - mouse 91%, rat 94%.

C21orf91 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf91
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C21orf91
Gene Accession No. Q9NYK6
Gene Alias C21orf14; C21orf38; C21orf91; chromosome 21 open reading frame 91; cold sore susceptibility gene 1; CSSG1; early undifferentiated retina and lens; EURL; Protein EURL homolog; YG81
Gene Symbols C21orf91
Host Species Rabbit
Immunogen Recombinant Human C21orf91. Recombinant protein control fragment (Product #RP-106214).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.