Learn More
Invitrogen™ C21orf91 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113812
Description
Immunogen sequence: LCRNSVLWPH SHNQAQKKEE TISSPEANVQ TQHPHYSREE LNSMTLGEVE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 86%.
C21orf91 is a protein coding gene.
Specifications
| C21orf91 | |
| Polyclonal | |
| Unconjugated | |
| C21orf91 | |
| C21orf14; C21orf38; C21orf91; chromosome 21 open reading frame 91; cold sore susceptibility gene 1; CSSG1; early undifferentiated retina and lens; EURL; Protein EURL homolog; YG81 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54149 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NYK6 | |
| C21orf91 | |
| Synthetic peptide directed towards the middle region of human C21orf91. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.