Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C22orf13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546259 Supplier Invitrogen™ Supplier No. PA546259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GHFIVLRGYN RATGCIFYNN PAYADPGMCS TSISNFEEAR TSYGTDEDIL Sequence homology: Cow: 100%; Dog: 100%; Goat: 77%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GUCD1
Gene Accession No. Q96NT3
Gene Alias 1110038D17Rik; C17H22orf13; C22orf13; C26H22orf13; CG13760 gene product [Drosophila melanogaster] homolog; guanylyl cyclase domain containing 1; guanylyl cyclase domain-containing protein 1; GUCD1; LLN4; protein GUCD1; protein GUCD1 pseudogene; Protein LLN4; uncharacterized protein C22orf13 homolog; wu:fi27d12; zgc:152874
Gene Symbols GUCD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C22orf13 (aa 185-234).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83606
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.