Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C22orf13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110881 Supplier Invitrogen™ Supplier No. PA5110881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MHHFGVRHRF CTQTLGVDKG YKNQSFYRKH FDTEETRVNQ LFAQAKACKV LVEKCTVSVK DIQAHLAQGH VAIVLVNSGV LHCDLCS Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 69%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf13
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GUCD1
Gene Accession No. Q96NT3
Gene Alias 1110038D17Rik; C17H22orf13; C22orf13; C26H22orf13; CG13760 gene product [Drosophila melanogaster] homolog; guanylyl cyclase domain containing 1; guanylyl cyclase domain-containing protein 1; GUCD1; LLN4; protein GUCD1; protein GUCD1 pseudogene; Protein LLN4; uncharacterized protein C22orf13 homolog; wu:fi27d12; zgc:152874
Gene Symbols GUCD1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human C22orf13. Recombinant protein control fragment (Product #RP-92360).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83606
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.