Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C22orf13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113200
Description
C22orf13 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The function of this protein remains unknown.
Specifications
C22orf13 | |
Polyclonal | |
Unconjugated | |
GUCD1 | |
1110038D17Rik; C17H22orf13; C22orf13; C26H22orf13; CG13760 gene product [Drosophila melanogaster] homolog; guanylyl cyclase domain containing 1; guanylyl cyclase domain-containing protein 1; GUCD1; LLN4; protein GUCD1; protein GUCD1 pseudogene; Protein LLN4; uncharacterized protein C22orf13 homolog; wu:fi27d12; zgc:152874 | |
Rabbit | |
Protein G | |
RUO | |
68778, 83606 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8BZI6, Q96NT3 | |
GUCD1 | |
Recombinant Human Protein GUCD1 protein (1-136AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction