Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C22orf13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983482
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3482 Supplier Bioss Supplier No. BS15131R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen C22orf13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GUCD1
Gene Accession No. Q96NT3
Gene Alias 1110038D17Rik; BOS_16819; C17H22orf13; C22orf13; C26H22orf13; CG13760 gene product [Drosophila melanogaster] homolog; chromosome 17 open reading frame, human C22orf13; chromosome 22 open reading frame 13; guanylyl cyclase domain containing 1; Guanylyl cyclase domain-containing protein 1; Gucd1; LLN4; LOC100357533; Protein GUCD1; protein GUCD1 pseudogene; Protein LLN4; uncharacterized protein C22orf13 homolog; wu:fi27d12; zgc:152874
Gene Symbols GUCD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C22orf13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83606
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.