Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C22orf23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581886
Description
Immunogen sequence: LQCSPTSSQR VLPSKQIASP IYLPPILAAR PHLRPANMCQ ANGAYSREQF KPQATRDLEK EKQRLQNIFA TGKDMEERKR KAPPARQKAP APELDRFEEL VKEIQERKEF LADMEALGQG KQYRGIILAE ISQKLREMED IDHRRSEE.
C22orf23 is a protein coding gene.
Specifications
C22orf23 | |
Polyclonal | |
Unconjugated | |
C22orf23 | |
C22orf23; chromosome 22 open reading frame 23; dJ1039K5.6; EVG1; UPF0193 protein EVG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZE7 | |
C22orf23 | |
Recombinant protein corresponding to Human C22orf23. Recombinant protein control fragment (Product #RP-91389). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction