Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C22orf28 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9500 Supplier Bioss Supplier No. BS9985R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen C22orf28
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rtcb
Gene Accession No. Q9Y3I0
Gene Alias AI255213; AI463255; ankyrin repeat domain 54; BOS_5645; C22orf28; C5H22orf28; chromosome 5 open reading frame, human C22orf28; D10Wsu52e; DJ149A16.6; FAAP; Focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein LOC406376; LOC100346526; p55; RNA 2′,3′-cyclic phosphate and 5′-OH ligase; rtcb; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; uncharacterized LOC100346526; zgc:76871
Gene Symbols Rtcb
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HSPC117/C22orf28.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51493
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.