Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C22orf29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551574 Supplier Invitrogen™ Supplier No. PA551574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRSLPAAMAT PAVSGSNSVS RSALFEQQLT KESTPGPKEP PVLPSSTCSS KPGPVEPASS QPEEAAPTPV PRLSESANPP AQRPDPAHPG GPKPQKTEEE VLETEGDQEV SLGTPQEVVE APETPGEPPL SP Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 32%.

C22orf29, located in mitochondrion, is involved in mitochondrial outer membrane permeabilization and regulation of mitochondrial membrane potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf29
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTL10
Gene Accession No. Q7L3V2
Gene Alias BH3-only protein; BOP; C22orf29; chromosome 22 open reading frame 29; protein Bop; Retrotransposon Gag-like protein 10; RTL10
Gene Symbols RTL10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C22orf29. Recombinant protein control fragment (Product #RP-91327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.