Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C22orf31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551423
Description
Immunogen sequence: LTVRQDLEDR YAEHVAATQA LPQDSGTAAW KGRVLLPETQ KRQQLSEDTL TIHGLPTEGY QALYHAVVEP MLWNPSGTPK RYSLELGKAI KQKLWEALCS QGAISEGAQR DRFPGRKQPG VHEEPVLKKW PKLKSK Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 25%.

Specifications
C22orf31 | |
Polyclonal | |
Unconjugated | |
C22orf31 | |
bK747E2.1; C22orf31; chromosome 22 open reading frame 31; HS747E2A; uncharacterized protein C22orf31 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25770 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95567 | |
C22orf31 | |
Recombinant protein corresponding to Human C22orf31. Recombinant protein control fragment (Product #RP-91401). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction