Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C22orf32 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983487
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3487 Supplier Bioss Supplier No. BS15136R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Essential regulatory subunit of the mitochondrial calcium uniporter complex (uniplex), a complex that mediates calcium uptake into mitochondria (PubMed:24231807, PubMed:26774479, PubMed:27099988). Required to bridge the calcium-sensing proteins MICU1 and MICU2 with the calcium-conducting subunit MCU (PubMed:24231807). Plays a central role in regulating the uniplex complex response to intracellular calcium signaling (PubMed:27099988). Acts by mediating activation of MCU and retention of MICU1 to the MCU pore, in order to ensure tight regulation of the uniplex complex and appropriate responses to intracellular calcium signaling (PubMed:27099988).

Specifications

Antigen C22orf32
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SMDT1
Gene Accession No. Q9H4I9
Gene Symbols SMDT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C22orf32.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91689
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.