Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C22orf36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544471 Supplier Invitrogen™ Supplier No. PA544471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WKSSDKICRQ LIYHLTPHSK QQQGSSLRQR KTQSCLKSSL QKTLLAGETV Sequence homology: Human: 100%; Rat: 100%.

The function of C22orf36 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf36
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC75B
Gene Accession No. Q2VPJ9
Gene Alias AI646023; C22orf36; Fam211b; family with sequence similarity 211, member B; leucine rich repeat containing 75B; leucine-rich repeat-containing protein 75B; leucine-rich repeat-containing protein C22orf36; leucine-rich repeat-containing protein C22orf36 homolog; leucine-rich repeat-containing protein FAM211B; LRRC75B
Gene Symbols LRRC75B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C22orf36.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388886
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.