Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C22orf39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568487 Supplier Thermo Scientific Supplier No. PA568487
Add to Cart
Add to Cart

C22orf39 Polyclonal Antibody

C22orf39 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf39
Applications Western Blot
Classification Polyclonal
Concentration 0.5 to 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C22orf39
Gene Accession No. Q6P5X5
Gene Alias C22orf39; chromosome 22 open reading frame 39; UPF0545 protein; UPF0545 protein C22orf39
Gene Symbols C22orf39
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C22orf39.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128977
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.