Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C22orf41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583880 Supplier Invitrogen™ Supplier No. PA583880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DADPEERNYD NMLKMLSDLN KDLEKLLEEM EKISVQATWM AYDMVVMRTN PTLAESMRRL EDAFVNCKEE MEKNWQELLH ETKQR Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 91%.

Major component of the transverse central element of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase. Required for chromosome loading of the central element-specific SCS proteins, and for initiating synapsis between homologous chromosomes. Chromosome loading appears to require SYCP1. Required for fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C22orf41
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYCE3
Gene Accession No. A1L190
Gene Alias C22orf41; SYCE3; Synaptonemal complex central element protein 3; testis highly expressed gene 2 protein; testis highly expressed protein 2; THEG2; THEG-2
Gene Symbols SYCE3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYCE3. Recombinant protein control fragment (Product #RP-100352).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644186
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.