Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ C25-140
SDP

Catalog No. 771510 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-151-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77-151-0 Supplier Tocris Bioscience™ Supplier No. 7715/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TRAF6-Ubc13 interaction inhibitor

C25-140 binds directly to TNF receptor-associated factor 6 (TRAF6) and inhibits its interaction with ubiquitin-conjugating enzyme Ubc13. C25-140 reduces TRAF6-Ubc13 activity and decreases NF-κB activation and inflammatory signaling in vitro. C25-140 exhibits selectivity for TRAF6 over other E3 ligases except cIAP1. In in vivo mouse models of autoimmune psoriasis and rheumatoid arthritis, the compound improves disease symptoms. The compound also suppresses the pro-inflammatory signaling activity of IL-17A and prevents its inhibitory effect on the anti-HBV action of IFN-α HepG2 cells.

Specifications

Chemical Name or Material 3-[3,5-Dimethyl-1-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)-1H-pyrazol-4-yl]-1-[4-(phenylmethyl)-1-piperidinyl]-1-propanone
CAS 1358099-18-9
Quantity 10 mg
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C26H31N7O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.