Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2CD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556592 Supplier Invitrogen™ Supplier No. PA556592
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKSKELHLQI SEAGRSSEGL LATATVPLDL FKKQPSGPQS FTLTSGSACG SSVLGSVTAE FSYMEPGELK SWPIP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

The C2CD2 gene, located on chromosome 2, encodes a protein that contains two C2 domains and is involved in cellular signaling and membrane trafficking. The protein has been implicated in various cellular processes, including calcium-dependent phospholipid binding and vesicle transport. Recent studies have shown a critical role for C2CD2 in the regulation of immune responses and cell apoptosis. The protein also interacts with other cellular components to modulate pathways that are essential for cell survival and proliferation. Mutations or dysregulation of C2CD2 have been linked to certain diseases, such as cancers and autoimmune disorders, highlighting its potential as a biomarker and therapeutic target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2CD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C2CD2
Gene Accession No. Q9Y426
Gene Alias 5730563M15; 5830404H04Rik; AI314146; AI449533; BF168443; C2 calcium dependent domain containing 2; C2 calcium-dependent domain containing 2; C2 domain-containing protein 2; C21orf25; C21orf258; C2cd2; Kiaa4045; mKIAA4045; ORF25; RGD735112; TMEM24 related geme; TMEM24L; Transmembrane protein 24-like
Gene Symbols C2CD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C2CD2. Recombinant protein control fragment (Product #RP-95881).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.