Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2CD3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA572860
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572860 Supplier Thermo Scientific Supplier No. PA572860

Rabbit Polyclonal Antibody

Specificity: C2CD3 antibody is human, mouse and rat reactive.

C2CD3 Polyclonal antibody specifically detects C2CD3 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2CD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene C2CD3
Gene Accession No. Q4AC94, Q52KB6
Gene Alias AU020772; C2 calcium dependent domain containing 3; C2 calcium-dependent domain containing 3; C2 domain-containing protein 3; C2CD3; Hty; OFD14; Protein hearty; RGD1307366
Gene Symbols C2CD3
Host Species Rabbit
Immunogen C2CD3 antibody for an 18 amino acid peptide near the center of human C2CD3. The immunogen is within amino acids 1490 - 1540 of C2CD3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26005, 277939, 293148
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.