Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2CD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547917
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547917 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547917 Supplier Thermo Scientific Supplier No. PA547917
Add to Cart
Add to Cart

C2CD3 Polyclonal Antibody

C2CD3 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen C2CD3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene C2CD3
Gene Accession No. Q4AC94
Gene Alias AU020772; C2 calcium dependent domain containing 3; C2 calcium-dependent domain containing 3; C2 domain-containing protein 3; C2CD3; Hty; OFD14; Protein hearty; RGD1307366
Gene Symbols C2CD3
Host Species Sheep
Immunogen E. coli-derived recombinant human C2CD3 Met1-Asp109.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.