Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2CD4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145196 Supplier Invitrogen™ Supplier No. PA5145196
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPPRLMPRLA LAALRNSWVE EAGMD Antigen sequence identity: Mouse 52%; Rat 52%.

Protein C2 calcium dependent domain containing 4A (C2CD4A) is a protein-coding gene that is involved in the regulation of calcium-dependent signaling pathways. It is expressed in various tissues and has been implicated in the development of certain cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2CD4A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C2CD4A
Gene Accession No. Q8NCU7
Gene Alias C2 domain-containing protein 4A; FAM148A
Gene Symbols C2CD4A
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 51-75. Recombinant protein control fragment (Product #RP-109945).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.