Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C2CD5 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074595AF647
Description
C2CD5 Polyclonal antibody specifically detects C2CD5 in Human samples. It is validated for Western BlotSpecifications
C2CD5 | |
Polyclonal | |
Western Blot | |
DKFZp779N2044, hypothetical protein LOC9847, KIAA0528 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human KIAA0528 using the numbering given in entry NP_055617.1 (GeneID 9847). | |
0.1 mL | |
Cell Biology, Signal Transduction | |
9847 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction