Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C2CD5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074596
Description
C2CD5 Polyclonal specifically detects C2CD5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
KIAA0528 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp779N2044, hypothetical protein LOC9847, KIAA0528 | |
Rabbit | |
Affinity Purified | |
RUO | |
9847 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
Q86YS7 | |
C2CD5 | |
The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human KIAA0528 using the numbering given in entry NP_055617.1 (GeneID 9847). | |
100 μL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Purple sea urchin, Florida lancelet and Spotted green pufferfish. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction