Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2CD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109466 Supplier Invitrogen™ Supplier No. PA5109466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSSLNLTNQ ALNKNFNDLC ENLLKSLYFK LRSMIPCCLC HVNFTVSLPE DELIQVTVTA VAITFDKNQA LQTTKTPVEK SLQRASTDNE ELLQFPLELC SDSLPSHPFP PAKEHLESAS SNSGIPAAQR ATSVDYSSFA DRCSSWIELI KLKAQTIRRG SIKTTMTVEK ASPVGDGNFR NRSAPPCANS TVGVVKMTPL SFIPGAKITK YLGIINMFFI RETTSLREEG GVSGFLHAFI AEVFAMVRAH VAALGGNAVV SYIMKQCVFM ENPNKNQAQC LINVSGDAVV FVRESDLEVV SSQQPTTNCQ SSCTEGEVTT.

C2CD5 gene ontology annotations related to this gene include calcium-dependent phospholipid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2CD5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene C2CD5
Gene Accession No. Q7TPS5, Q86YS7
Gene Alias 138 kDa C2 domain-containing phosphoprotein; 5730419I09Rik; C030008B15Rik; C2 calcium dependent domain containing 5; C2 calcium-dependent domain containing 5; C2 domain-containing phosphoprotein of 138 kDa; C2 domain-containing protein 5; C2CD5; Cdp138; KIAA0528; RGD1304592
Gene Symbols C2CD5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 732-1051 of human C2CD5 (NP_001273103.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362461, 74741, 9847
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.