Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543026 Supplier Thermo Scientific Supplier No. PA543026
Add to Cart
Add to Cart

C2orf18 Polyclonal Antibody

C2orf18 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC35F6
Gene Accession No. Q8N357
Gene Alias 4930471M23Rik; 5730478O19Rik; ANT2-binding protein; ANT2BP; C11H2orf18; C17H2orf18; C2orf18; CB018; P40; RGD1309228; sb:cb433; SLC35F6; Solute carrier family 35 member F6; solute carrier family 35, member F6; TANGO9; transmembrane protein C2orf18; transmembrane protein C2orf18 homolog; transport and Golgi organization 9 homolog; UNQ3047/PRO9863; wu:fd19b11; wu:fi67c08; zgc:92765
Gene Symbols SLC35F6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C2orf18.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54978
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.