Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C2orf18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557003
Description
Immunogen sequence: GLHRPLLGRL SRGRPLAEES EQERLLGGTR TPINDAS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.
The exact function of C2orf18 remains unknown. It contains 1 DUF6 domain.
Specifications
| C2orf18 | |
| Polyclonal | |
| Unconjugated | |
| SLC35F6 | |
| 4930471M23Rik; 5730478O19Rik; ANT2-binding protein; ANT2BP; C11H2orf18; C17H2orf18; C2orf18; CB018; P40; RGD1309228; sb:cb433; SLC35F6; Solute carrier family 35 member F6; solute carrier family 35, member F6; TANGO9; transmembrane protein C2orf18; transmembrane protein C2orf18 homolog; transport and Golgi organization 9 homolog; UNQ3047/PRO9863; wu:fd19b11; wu:fi67c08; zgc:92765 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N357 | |
| SLC35F6 | |
| Recombinant protein corresponding to Human C2orf18. Recombinant protein control fragment (Product #RP-96244). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction