Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989359
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9359 Supplier Bioss Supplier No. BS9804R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CNPPD1 gene ontology annotations related to this gene include regulation of cyclin-dependent protein serine/threonine kinase activity.

Specifications

Antigen C2orf24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CNPPD1
Gene Accession No. Q9BV87
Gene Alias 1810031K17Rik; AU043960; BOS_2094; C2H2orf24; C2orf24; CDABP0125; CGI-57; Chromosome 2 open reading frame 24 ortholog-like; CNPPD1; cyclin Pas1/PHO80 domain containing 1; Cyclin Pas1/PHO80 domain-containing protein 1; hypothetical protein MGC:72616; LOC100353345; LOC27013; Protein CNPPD1; RGD735175; Similar to hypothetical protein CGI-57; wu:fa05h07; zgc:55857
Gene Symbols CNPPD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C2orf24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27013
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.