Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf27B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen C2orf27B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156812
SDP
View Documents
Novus Biologicals
NBP156812
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

C2orf27B Polyclonal specifically detects C2orf27B in Human samples. It is validated for Western Blot.
Specifications

Specifications

C2orf27B
Polyclonal
Rabbit
Human
chromosome 2 open reading frame 27B, hypothetical protein LOC408029, MGC50273
C2ORF27B
IgG
Western Blot
Unconjugated
RUO
Q580R0-2
408029
Synthetic peptides corresponding to MGC50273(MGC50273 protein) The peptide sequence was selected from the N terminal of MGC50273. Peptide sequence MFVRPESGEQGPETLAPASGAEIQRFPVPAVEPVPAPGADSPPGTALELE.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.