Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf28 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989361
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9361 Supplier Bioss Supplier No. BS9806R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.

Specifications

Antigen C2orf28
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATRAID
Gene Accession No. Q6UW56
Gene Alias 0610007C21Rik; 3-Apr; AI316792; all-trans retinoic acid induced differentiation factor; All-trans retinoic acid-induced differentiation factor; apoptosis related protein 3; apoptosis related protein 3-like; apoptosis related protein APR-3; Apoptosis-related protein 3; APR3; APR-3; APR--3; ATRAID; BOS_11795; C11H2orf28; C17H2orf28; C2orf28; chromosome 11 open reading frame, human C2orf28; H1E6 protein; HSPC013; LOC100352483; p18; p18 protein; PRO240; RGD1311605; UNQ214/PRO240
Gene Symbols ATRAID
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C2orf28 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51374
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.