Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114127 Supplier Invitrogen™ Supplier No. PA5114127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEPDDHEKRI PFTLRPQTLR QRMAEESISR NEETSEESQE DEKQDTWEQQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 100%; Guinea Pig: 92%; Horse: 85%; Human: 100%; Mouse: 85%; Rabbit: 83%; Rat: 92%.

TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GCFC2
Gene Accession No. P16383
Gene Alias A130099G21; AW146020; C2orf3; DNABF; GC binding factor; GC bindng factor; GCF; GCF2; GCFC2; GC-rich sequence DNA binding factor 2; GC-rich sequence DNA-binding factor; GC-rich sequence DNA-binding factor 2; RGD1304792; TCF9; TCF-9; Transcription factor 9; transcription factor 9 (binds GC-rich sequences)
Gene Symbols GCFC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human C2orf3 (aa 180-229).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6936
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.