Learn More
Invitrogen™ C2orf33 Polyclonal Antibody

Description
Immunogen sequence: DLIQSTPFKPL ALKTPPRVLT LSERPLDFLD LERPPTTPQN EEIRAVGRLK RERSMSENAV RQNGQLVRND SLWH Highest antigen sequence identity to the following orthologs - mouse 93%, rat 96%.
Specifications
Specifications
| Antigen | C2orf33 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MFF |
| Gene Accession No. | Q9GZY8 |
| Gene Alias | 5230400G24Rik; AD030; AD033; AI314724; C2H2orf33; C2orf33; GL004; hypothetical protein LOC393508; hypothetical protein LOC506291; MFF; mffa; Mitochondrial fission factor; mitochondrial fission factor a; mitochondrial fission factor homolog B; RGD1310230; zgc:66022 |
| Gene Symbols | MFF |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.