Learn More
Invitrogen™ C2orf33 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592310
Description
Positive Samples: HeLa; Cellular Location: Cytoplasmic vesicle, Mitochondrion outer membrane, Peroxisome, Single-pass type IV membrane protein, secretory vesicle, synaptic vesicle Immunogen sequence: MSKGTSSDTS LGRVSRAAFP SPTAAEMAEI SRIQYEMEYT EGISQRMRVP EKLKVAPPNA DLEQGFQEGV PNASVIMQVP ERIVVAGNNE DVSFSRPADL.
Plays a role in mitochondrial and peroxisomal fission. Promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface. May be involved in regulation of synaptic vesicle membrane dynamics by recruitment of DNM1L to clathrin-containing vesicles. [Uniprot].
Specifications
C2orf33 | |
Polyclonal | |
Unconjugated | |
MFF | |
5230400G24Rik; AD030; AD033; AI314724; C2H2orf33; C2orf33; GL004; hypothetical protein LOC393508; hypothetical protein LOC506291; MFF; mffa; Mitochondrial fission factor; mitochondrial fission factor a; mitochondrial fission factor homolog B; RGD1310230; zgc:66022 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
301563, 56947, 75734 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q4KM98, Q6PCP5, Q9GZY8 | |
MFF | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MFF (NP_0645793). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.