Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf42 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen C2orf42
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170466
SDP
View Documents
Novus Biologicals
NBP170466
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

C2orf42 Polyclonal specifically detects C2orf42 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C2orf42
Polyclonal
Rabbit
chromosome 2 open reading frame 42
C2ORF42
IgG
63 kDa
Western Blot
Unconjugated
RUO
54980
Synthetic peptides corresponding to C2ORF42 The peptide sequence was selected from the N terminal of C2ORF42. Peptide sequence EPNSLRTKVPAFLSDLGKATLRGIRKCPRCGTYNGTRGLSCKNKTCGTIF.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.