Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C2orf43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113410
Description
C2orf43 Polyclonal Antibody for Western Blot, IHC (P), ELISA
C2orf43 is a protein coding gene.
Specifications
| C2orf43 | |
| Polyclonal | |
| Unconjugated | |
| LDAH | |
| 1110057K04Rik; C2orf43; hLDAH; LDAH; LD-associated hydrolase; lipid droplet associated hydrolase; lipid droplet-associated hydrolase; Lipid droplet-associated serine hydrolase; mLDAH; RGD1311648; similar to hypothetical protein FLJ21820; UPF0554 protein C2orf43; UPF0554 protein C2orf43 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60526 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H6V9 | |
| LDAH | |
| Recombinant Human Lipid droplet-associated hydrolase protein (1-325AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction