Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584889 Supplier Invitrogen™ Supplier No. PA584889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAHVSKLLSQ CKFDLLEELV AKEVLHALKE KVTSLPDNHK NALAANIDEI V Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 86%.

Promotes sorting of SMDT1/EMRE in mitochondria by ensuring its maturation. Interacts with the transit peptide region of SMDT1/EMRE precursor protein in the mitochondrial matrix, leading to protect it against protein degradation by YME1L1, thereby ensuring SMDT1/EMRE maturation by the mitochondrial processing peptidase (PMPCA and PMPCB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf47
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAIP1
Gene Accession No. Q8WWC4
Gene Alias 9430016H08Rik; AA409999; C2orf47; C78988; chromosome 2 open reading frame 47; m-AAA protease-interacting protein 1, mitochondrial; Maip1; Matrix AAA peptidase-interacting protein 1; MGC94335; RIKEN cDNA 9430016H08 gene; similar to hypothetical protein FLJ22555; uncharacterized protein C2orf47 homolog, mitochondrial; uncharacterized protein C2orf47, mitochondrial
Gene Symbols MAIP1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human MAIP1 (Product #RP-105297)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79568
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.