Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570939 Supplier Invitrogen™ Supplier No. PA570939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 93%; Guinea Pig: 77%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 93%.

C2orf54 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf54
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C2orf54
Gene Accession No. Q08AI8
Gene Alias C2orf54; chromosome 2 open reading frame 54; MAB21L4; Protein mab-21-like 4; uncharacterized protein C2orf54
Gene Symbols C2orf54
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human C2orf54.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79919
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.