Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C2orf57 Polyclonal Antibody

C2orf57 Polyclonal Antibody
Supplier: Thermo Scientific PA563302
Description
C2orf57 Polyclonal Antibody for IHC
Specifications
| C2orf57 | |
| Polyclonal | |
| Unconjugated | |
| TEX44 | |
| C2orf57; chromosome 2 open reading frame 57; Testis-expressed protein 44; TEX44; uncharacterized protein C2orf57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 165100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53QW1 | |
| TEX44 | |
| Recombinant protein corresponding to Human C2orf57. Recombinant protein control fragment (Product #RP-99692). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction