Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf57 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989371
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9371 Supplier Bioss Supplier No. BS9818R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C2orf57
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TEX44
Gene Accession No. Q53QW1
Gene Alias C2orf57; chromosome 2 open reading frame 57; Testis-expressed protein 44; TEX44; uncharacterized protein C2orf57
Gene Symbols TEX44
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C2orf57.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165100
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.