Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf60 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565020 Supplier Invitrogen™ Supplier No. PA565020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRVVLFSPRDA QYLYLKGTKS EVLNIDNPDL AKYPLFSKAR RYECSLEAGD VLFIPALWFH NVISEEFGVG VNIFWKHLPS ECYDKTDTYG NKDPTAASRA AQILDRALKT LAELPEEYRD FYARRMV Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.

tRNA hydroxylase that acts as a component of the wybutosine biosynthesis pathway. Wybutosine is a hyper modified guanosine with a tricyclic base found at the 3'-position adjacent to the anticodon of eukaryotic phenylalanine tRNA. Catalyzes the hydroxylation of 7-(a-amino-a-carboxypropyl)wyosine (yW-72) into undermodified hydroxywybutosine (OHyW*). OHyW* being further transformed into hydroxywybutosine (OHyW) by LCMT2/TYW4. OHyW is a derivative of wybutosine found in higher eukaryotes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf60
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TYW5
Gene Accession No. A2RUC4
Gene Alias C2orf60; hTYW5; jmjC domain-containing protein C2orf60; tRNA wybutosine-synthesizing protein 5; tRNA yW-synthesizing enzyme 5; tRNA(Phe) (7-(3-amino-3-carboxypropyl)wyosine(37)-C(2))-hydroxylase; tRNA-yW synthesizing protein 5; TYW5
Gene Symbols TYW5
Host Species Rabbit
Immunogen Recombinant Human C2orf60. Recombinant protein control fragment (Product #RP-89696).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129450
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.