Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562666 Supplier Thermo Scientific Supplier No. PA562666
Add to Cart
Add to Cart

C2orf61 Polyclonal Antibody

C2orf61 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf61
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STPG4
Gene Accession No. Q8N801
Gene Alias 1700011E24Rik; C2orf61; chromosome 2 open reading frame 61; Gonad-specific expression gene protein; GSE; hypothetical protein LOC681766; LOC681766; Protein STPG4; RIKEN cDNA 1700011E24 gene; Sperm-tail PG-rich repeat-containing protein 4; STPG4; uncharacterized protein C2orf61; uncharacterized protein C2orf61 homolog
Gene Symbols STPG4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C2orf61. Recombinant protein control fragment (Product #RP-103132).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285051
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.