Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C2orf62 Monoclonal Antibody (OTI2F4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505502S


Description
C2orf62 Monoclonal antibody specifically detects C2orf62 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
C2orf62 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y296 | |
Mouse | |
Affinity chromatography | |
RUO | |
51399 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2F4 | |
Unconjugated | |
Trappc4 | |
C15H2orf62; C2orf62; C37H2orf62; C6H2orf62; Catip; ciliogenesis associated TTC17 interacting protein; Ciliogenesis-associated TTC17-interacting protein; Gm216; hypothetical protein LOC690437; LOC690437 | |
Full length human recombinant protein of human TRAPPC4 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction