Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C2orf68 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | C2orf68 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
C2orf68 Polyclonal specifically detects C2orf68 in Human samples. It is validated for Western Blot.Specifications
C2orf68 | |
Polyclonal | |
Rabbit | |
chromosome 2 open reading frame 68, FLJ35653, MGC131675 | |
C2orf68 | |
IgG | |
19 kDa |
Western Blot | |
Unconjugated | |
RUO | |
388969 | |
Synthetic peptides corresponding to LOC388969 The peptide sequence was selected from the N terminal of LOC388969. Peptide sequence VRRRHTPAPTRPRKPDLQVYLPRHRDVSAHPRNPDYEESGESSSSGGSEL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title