Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571025 Supplier Invitrogen™ Supplier No. PA571025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 93%; Human: 100%; Rabbit: 86%.

C2orf70 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf70
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C2orf70
Gene Accession No. A6NJV1
Gene Alias C2orf70; chromosome 2 open reading frame 70; FAM166C; Family with sequence similarity 166 member C; Protein FAM166C; UPF0573 protein; UPF0573 protein C2orf70
Gene Symbols C2orf70
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human C2orf70.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339778
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.