Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C2orf78 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145053
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145053 Supplier Invitrogen™ Supplier No. PA5145053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQNTSLPGTA NSRQFSLPVV SNAAFLTGSI SNFSRASAPA ISSAWLQPSA SGTSFQPLMG SAYLYQHSST TMLSGVTG Antigen sequence identity: Mouse 51%; Rat 53%.

C2orf78 is a protein that is encoded by the C2orf78 gene in humans. It is a poorly characterized protein with unknown function and has been found to be expressed in various tissues including the brain, liver, and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf78
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C2orf78
Gene Accession No. A6NCI8
Gene Alias Chromosome 2 Open Reading Frame 78; HCG1989538; Uncharacterized Protein C2orf78
Gene Symbols C2orf78
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 35-112. Recombinant protein control fragment (Product #RP-110020).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.