Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf80 Mouse anti-Human, Clone: OTI3B6, liquid, TrueMAB™
SDP

Catalog No. 501682142
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2142 Supplier OriGene Supplier No. TA808450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C2orf80 (Chromosome 2 Open Reading Frame 80) is a Protein Coding gene. Diseases associated with C2orf80 include Gonadal Dysgenesis and 46 Xy Gonadal Dysgenesis.

Specifications

Antigen C2orf80
Applications Western Blot
Classification Monoclonal
Clone OTI3B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C2orf80
Gene Accession No. Q0P641
Gene Alias GONDA1
Gene Symbols C2orf80
Host Species Mouse
Immunogen Full length human recombit protein of human C2orf80 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389073
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.