Learn More
Invitrogen™ C2orf82 Polyclonal Antibody

Description
Immunogen sequence: TDDVPQEPVP TLWNEPAELP SGEGPVESTS PGREPVDTGP PAPTVAPGPE DSTAQE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 40%.
Specifications
Specifications
| Antigen | C2orf82 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C2orf82 |
| Gene Accession No. | Q6UX34 |
| Gene Alias | ASCL830; C2orf82; chromosome 2 open reading frame 82; Protein SNORC; Secondary ossification center-associated regulator of chondrocyte maturation protein; SNORC; uncharacterized protein C2orf82; UNQ830; UNQ830/PRO1757 |
| Gene Symbols | C2orf82 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.